Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cerebellin-4 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF6740SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6740SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6740SP Supplier R&D Systems Supplier No. AF6740SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Cerebellin-4 Polyclonal specifically detects Cerebellin-4 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Cerebellin-4
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NTU7
Gene Alias CBLN4, CBLNL1, cerebellin 4 precursor, cerebellin precursor-like 1, cerebellin-4, Cerebellin4, Cerebellin-like glycoprotein 1, dJ885A10.1
Gene Symbols CBLN4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Cerebellin-4 Gln28-Leu201 Accession # Q9NTU7
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140689
Test Specificity Detects human Cerebellin-4 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Cerebellin-1 is observed, and less than 5% cross-reactivity with rhCerebellin-2 and rhCerebellin-3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.