Learn More
Sino Biological Human CFH PAb 100UL 10714-RP02-100
Supplier:  Sino Biological 10714RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CFH (rh CFH; Catalog#10714-H08H; NP_000177.2; Ser 860-Arg 1231). CFH specific IgG was purified by human CFH affinity chromatography.
Catalog No. 50-241-0459
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.