Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Chemerin Recombinant Protein, PeproTech®

Catalog No. 300661MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-66-1MG 1 mg
30066100UG 100 μg
30066250UG 250 μg
300-66-25UG 25 μg
30066500UG 500 μg
300-66-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 300-66-1MG Supplier Gibco™ Supplier No. 300661MG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Chemerin is a 15.6 kDa protein consisting of 135 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Retinoids act through ligand-dependent transcription factors, retinoid X receptor (RXRs) and retinoic acid receptors (RARs). Tazarotene-induced gene (TIG) proteins, also designated retinoic acid receptor responder proteins or RAR-responsive proteins, can be membrane bound or secreted. TIGs act as tumor suppressor genes in human cancers and are highly expressed in skin, hair follicles and endothelial cells as well as in pancreas, spleen and intestine. TIGs are activated by tazarotene and have been implicated as growth regulators that mediate the growth suppressive effects of retinoids. TIG2 is a secreted protein that is mainly expressed in epidermis, hair follicles and endothelial cells. TIG2 is inhibited in psoriatic lesions and is activated by tazarotene in skin rafts and in epidermis of psoriatic lesions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99969
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 5919
Molecular Weight (g/mol) 15.6 kDa
Name Human Chemerin
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 0610007L05Rik; AI303516; Chemerin; HP10433; Rarres2; RAR-responsive protein TIG2; retinoic acid receptor responder (tazarotene induced) 2; retinoic acid receptor responder 2; retinoic acid receptor responder protein 2; Tazarotene-induced gene 2 protein; TIG2; TIG-2; TIG2HP10433
Common Name Chemerin
Gene Symbol Rarres2
Biological Activity Determined by its ability to chemoattract human immature dendritic cells using a concentration range of 1.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ELTEAQRRGL QVALEEFHKH PPVQWAFQET SVESAVDTPF PAGIFVRLEF KLQQTSCRKR DWKKPECKVR PNGRKRKCLA CIKLGSEDKV LGRLVHCPIE TQVLREAEEH QETQCLRVQR AGEDPHSFYF PGQFA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.