Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Chitinase 3-like 1/YKL-40 Quantikine ELISA Kit
SDP

Catalog No. DC3L10
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DC3L10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DC3L10 Supplier R&D Systems™ Supplier No. DC3L10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Chitinase 3-like 1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Chitinase 3-like 1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Chitinase 3-like 1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).

Specifications

Assay Range 62.50 to 4000 pg/mL
Assay Sensitivity 8.15 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human CHI3L1
For Use With (Application) ELISA
Gene ID (Entrez) 1116
Gene Symbol CHI3L1
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40
Target Chitinase 3-like 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.