Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CHL-1/L1CAM-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2126
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2126 100 μg
MAB2126SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2126 Supplier R&D Systems Supplier No. MAB2126
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

CHL-1/L1CAM-2 Monoclonal specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CHL-1/L1CAM-2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 316223
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1.7-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_006605
Gene Alias CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Gene Symbols CHL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CHL-1/L1CAM-2 Ile25-Gln1096 Accession # NP_006605
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10752
Test Specificity Detects human CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CHL-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.