Learn More
Biomatik Corporation Human Chondroitin Sulfate Proteoglycan 4 (CSPG4) ELISA Kit
Supplier:  Biomatik Corporation EKU05891
Encompass_Preferred
Human Chondroitin Sulfate Proteoglycan 4 (CSPG4) ELISA Kit for detecting CSPG4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6UVK1. Gene ID: 1464. Target Synonyms: CSPG4; MSK16; HMW-MAA; MCSP; MCSPG; MEL-CSPG; NG2; Chondroitin Sulfate Proteoglycan 4; Melanoma chondroitin sulfate proteoglycan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8878
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.