Learn More
Human Chorionic Gonadotropin alpha/beta (HCG) Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | hCG |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1270D |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P0DN86 nad P0DN87 |
| Host Species | Rabbit |
| Immunogen | Human embryonic kidney cell HEK293-derived transfected with human Chorionic Gonadotropin beta , Accession # P0DN86 nad P0DN87 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.