Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human cIAP-1/BIRC2/hIAP-2 ELISA Kit

Catalog No. EH108RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH108RB 96 Tests
EH108RBX5 5 x 96 Tests
EH108RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH108RB Supplier Invitrogen™ Supplier No. EH108RB
Add to Cart
Add to Cart

ELISA

Human cIAP-1/BIRC2/hIAP-2 quantitates human cIAP-1/BIRC2/hIAP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human cIAP-1/BIRC2/hIAP-2.

The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number API1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW146227; baculoviral IAP repeat containing 2; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 2; BIRC2; Birc3; Cellular inhibitor of apoptosis 1; cIAP1; c-IAP1; cIAP2; HIAP1; HIAP2; Hiap-2; IAP homolog B; IAP1; IAP2; IAP-2; inhibitor of apoptosis protein; inhibitor of apoptosis protein 2; mcIAP1; MIAP1; MIAP2; mIAP-2; MIHB; MIHC; NFR2-TRAF signalling complex protein; rIAP1; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; RNF48; TNFR2-TRAF-signaling complex protein 2
Assay Range 1.65 to 400 ng/mL
Assay Sensitivity 1.65 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 329
Gene Symbol BIRC2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target cIAP1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.