Learn More
ACROBiosystems Human Claudin-1 500ug
Supplier:  ACROBiosystems CL1H52P6500UG
Encompass_Preferred
Human Claudin-1 Full Length Protein (VLP) (CL1-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 211 (Accession # O95832).
Catalog No. 50-253-5256
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.