Learn More
ACROBiosystems Human Claudin-6 / CLDN6 Protein, His,Twin-Strep Tag (Detergent)
Supplier:  ACROBiosystems CL6H5289500UG
Encompass_Preferred
Human Claudin-6 Protein, His,Twin-Strep Tag (CL6-H5289) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Val 220 (Accession # P56747-1).
Catalog No. 50-233-1459
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.