Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CLCNKB Full-length ORF (AAH20873.1, 1 a.a. - 462 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89939731 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-731 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-939-731 Supplier Abnova™ Supplier No. H00001188P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney. [provided by RefSeq]

Sequence: MEEFVGLREGSSGNPVTLQELWGPCPRIRRGIRGGLEWLKQKLFRLGEDWYFLMTLGVLMALVSCAMDLAVESVVRAHQWLYREIGDSHLLRYLSWTVYPVALVSFSSGFSQSITPSSGGSGIPEVKTMLAGVVLEDYLDIKNFGAKVVGLSCTLACGSTLFLGKVGPFVHLSVMMAAYLGRVRTTTIGEPENKSKPVYSALATLVLASITYPPSAGRFLASRLSMKQHLDSLFDNHSWALMTQNSSPPWPEELDPQHLWWEWYHPRFTIFGTLAFFLVMKFWMLILATTIPMPAGYFMPIFVYGAAIGRLFGETLSFIFPEGIVAGGITNPIMPGGYALAGAAAFSGAVTHTISTALLAFEVTGQIVHALPVLMAVLAANAIAQSCQPSFYDGTVIVKKLPYLPRILGRNIGSHRVRVEHFMNHSITTLAITLMNDEIGVHCHQGQAQMASGVVWFPVRGS

Specifications

Accession Number AAH20873.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1188
Molecular Weight (g/mol) 77kDa
Name CLCNKB (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MEEFVGLREGSSGNPVTLQELWGPCPRIRRGIRGGLEWLKQKLFRLGEDWYFLMTLGVLMALVSCAMDLAVESVVRAHQWLYREIGDSHLLRYLSWTVYPVALVSFSSGFSQSITPSSGGSGIPEVKTMLAGVVLEDYLDIKNFGAKVVGLSCTLACGSTLFLGKVGPFVHLSVMMAAYLGRVRTTTIGEPENKSKPVYSALATLVLASITYPPSAGRFLASRLSMKQHLDSLFDNHSWALMTQNSSPPWPEELDPQHLWWEWYHPRFTIFGTLAFFLVMKFWMLILATTIPMPAGYFMPIFVYGAAIGRLFGETLSFIFPEGIVAGGITNPIMPGGYALAGAAAFSGAVTHTISTALLAFEVTGQIVHALPVLMAVLAANAIAQSCQPSFYDGTVIVKKLPYLPRILGRNIGSHRVRVEHFMNHSITTLAITLMNDEIGVHCHQGQAQMASGVVWFPVRGS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CLCKB/ClC-K2/MGC24087/hClC-Kb
Common Name CLCNKB
Gene Symbol CLCNKB
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.