Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CLDN11 Full-length ORF (NP_005593.2, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89939740 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-740 25 ug
89-939-739 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-939-740 Supplier Abnova™ Supplier No. H00005010P01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene belongs to the claudin family of tight junction associated proteins and is a major component of central nervous system myelin that is necessary for normal CNS function. There is growing evidence that the protein determines the permeability between layers of myelin sheaths via focal adhesion and, with its expression highly regulated during development, may play an important role in cellular proliferation and migration. In addition, the protein is a candidate autoantigen in the development of autoimmune demyelinating disease. [provided by RefSeq]

Sequence: MVATCLQVVGFVTSFVGWIGVIVTTSTNDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACRALMIAASVLGLPAILLLLTVLPCIRMGQEPGVAKYRRAQLAGVLLILLALCALVATIWFPVCAHRETTIVSFGYSLYAGWIGAVLCLVGGCVILCCAGDAQAFGENRFYYTAGSSSPTHAKSAHV

Specifications

Accession Number NP_005593.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5010
Molecular Weight (g/mol) 48.4kDa
Name CLDN11 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MVATCLQVVGFVTSFVGWIGVIVTTSTNDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACRALMIAASVLGLPAILLLLTVLPCIRMGQEPGVAKYRRAQLAGVLLILLALCALVATIWFPVCAHRETTIVSFGYSLYAGWIGAVLCLVGGCVILCCAGDAQAFGENRFYYTAGSSSPTHAKSAHV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias OSP/OTM
Common Name CLDN11
Gene Symbol CLDN11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.