Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CLDN7 Full-length ORF (NP_001298.2, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89939775 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-775 10 ug
89-939-776 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-939-775 Supplier Abnova™ Supplier No. H00001366P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Claudins, such as CLDN7, are involved in the formation of tight junctions between epithelial cells. Tight junctions restrict lateral diffusion of lipids and membrane proteins, and thereby physically define the border between the apical and basolateral compartments of epithelial cells (Zheng et al., 2003 [PubMed 14502431]).[supplied by OMIM]

Sequence: MANSGLQLLGFSMALLGWVGLVACTAIPQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATRALMVVSLVLGFLAMFVATMGMKCTRCGGDDKVKKARIAMGGGIIFIVAGLATLVACSWYGHQIVTDFYNPLIPTNIKYEFGPAIFIGWAGSALVILGGALLSCSCPGNESKAGYRAPRSYPKSNSSKEYV

Specifications

Accession Number NP_001298.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1366
Molecular Weight (g/mol) 48.8kDa
Name CLDN7 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MANSGLQLLGFSMALLGWVGLVACTAIPQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATRALMVVSLVLGFLAMFVATMGMKCTRCGGDDKVKKARIAMGGGIIFIVAGLATLVACSWYGHQIVTDFYNPLIPTNIKYEFGPAIFIGWAGSALVILGGALLSCSCPGNESKAGYRAPRSYPKSNSSKEYV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CEPTRL2/CPETRL2/Hs.84359/claudin-1
Common Name CLDN7
Gene Symbol CLDN7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.