Learn More
ACROBiosystems Human CLEC12A 25tests
Supplier:  ACROBiosystems CLAHA24425TESTS
Encompass_Preferred
APC-Labeled Human CLEC12A Protein His Tag (CLA-HA244) is produced via conjugation of APC to Human CLEC12A Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CLEC12A Protein His Tag is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).
Catalog No. 50-253-5239
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.