Learn More
Description
- Also known as: Gop-1; KIAA0350
- CLEC16A is a member of the Ctype lectin/C-type lectin-like domain (CTL/CTLD) superfamily
- Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signaling, glycoprotein turnover, and roles in inflammation and immune response
- Single nucleotide polymorphisms in introns of CLEC16A have been associated with diabetes mellitus, multiple sclerosis and rheumatoid arthritis
- Alternatively spliced transcripts encoding different proteins have been described
- The CLEC16A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hCLEC16A |
| Accession Number | NP_056041 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3175 bp |
| Gene ID (Entrez) | 23274 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
