Learn More
iQ Biosciences Human Chronic Lymphocytic Leukemia PBMCs, 100M cells/vial
Supplier:  iQ Biosciences IQBPBMC203
Encompass_Preferred
High-quality, high-viability (> 90%) peripheral blood mononuclear cells (PBMCs) from chronic lymphocytic leukemia (CLL) blood donors sourced from a world-renowned blood center. Used for a wide variety of immunology-based applications, including in vitro immunoassays, ex vivo applications, and biologic testing. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate the number of vials from each individual donor - Donor(s) with a higher percentage of the cells of interest (e.g., B cells, CD4+ cells, CD8+ cells, NK cells, etc.) - Gender, age, body mass index (BMI), or ethnicity
Catalog No. NC2855601
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.