Learn More
R&D Systems™ Human CMG-2 DuoSet ELISA
Description
This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human CMG-2. The suggested diluent is suitable for the analysis of most cell culture supernate samples. Diluents for complex matrices, such as serum and plasma, should be evaluated prior to use in this DuoSet ELISA.
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
- Economical alternative to complete kits
Specifications
Specifications
| Assay Range | 187.00 to 12000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 118429 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | anthrax toxin receptor 2, ANTXR2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, cI-35, CMG2, CMG2MGC111533, CMG-2 mgC45856, FLJ31074, ISH, JHF, JHS |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.