Learn More
ACROBiosystems Human CNR2 500ug
Supplier:  ACROBiosystems CN2H52P5500UG
Encompass_Preferred
Human CNR2 Full Length Protein (VLP) (CN2-H52P5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Cys 360 (Accession # P34972-1).
Catalog No. 50-253-5234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.