Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coagulation Factor V (Light Chain) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9856SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9856SP 25 μg
MAB9856100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9856SP Supplier R&D Systems Supplier No. MAB9856SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Factor V Monoclonal specifically detects Factor V in Human samples. It is validated for ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen FactorV
Applications ELISA, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2280A
Conjugate Unconjugated
Dilution ELISA, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P12259
Gene Alias Activated protein C cofactor, Cf5, Cf-5, Coagulation Factor 5, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, F5, Factor V, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor, RPRGL1, THPH2
Host Species Rabbit
Immunogen Synthetic peptide containing human Coagulation Factor V aa1650-1700, Accession # P12259
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2153
Test Specificity Detects human Coagulation Factor V in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.