Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coagulation Factor X Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Catalog No. AF1063U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1063U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF1063U Supplier R&D Systems Supplier No. AF1063U
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Coagulation Factor X Polyclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen Coagulation Factor X
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation Supplied 0.2mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias Cf10, coagulation factor X, EC 3.4.21, EC 3.4.21.6, F10, factor Xa, FX, FXa, prothrombinase, Stuart Factor, Stuart-Prower factor
Host Species Goat
Immunogen S. frugiperda insect cell line Sf 21-derived recombinant human Coagulation Factor X, Leu24-Lys488, Accession # P00742
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2159
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.