Learn More
Description
- Also known as: FX; FXA
- Factor X is a vitamin K-dependent coagulation factor X of the blood coagulation cascade.
- This factor undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR.
- The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrinsic pathway), or by factor VIIa (in the extrinsic pathway).
- The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting.
- Mutations of Factor X result in factor X deficiency, a hemorrhagic condition of variable severity.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hF10 |
| Accession Number | NP_000495 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1480 bp |
| Gene ID (Entrez) | 2159 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
