Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human COCO ELISA Kit

Catalog No. EH120RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH120RB 96 Tests
EH120RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH120RB Supplier Invitrogen™ Supplier No. EH120RB
Add to Cart
Add to Cart

ELISA

Human COCO quantitates human COCO in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human COCO.

This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to bind Nodal and to inhibit the Nodal signaling pathway which patterns left/right body asymmetry.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CER2; CER2CRL2; cerberus 2; Cerberus like-2; cerberus-like 2; cerberus-like protein 2; CERL2; Cerl-2; Cerr2; CKTSF1B3; CKTSF1B3MGC126849; COCO; CRL2; Cysteine knot superfamily 1, BMP antagonist 3; DAN domain BMP antagonist family member 5; DAN domain family member 5; DAN domain family member 5, BMP antagonist; DAN domain family, member 5; DAND5; DANTE; Dte; FLJ38607; GREM3; GREM3cerberus 2; Gremlin-3; MGC126849; SP1
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 199699
Gene Symbol DAND5
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target DAND5
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.