Learn More
Novus Biologicals™ Human Collagen IV ELISA Kit (Chemiluminescence)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Assay Range | 0.16-10 ng/mL |
| Assay Sensitivity | 0.09 ng/mL |
| Cross Reactivity | This kit recognizes Human COL4 in samples. No significant cross-reactivity or interference between Human COL4 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Collagen IV |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.