Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Collagen X ELISA Kit (Chemiluminescence)
Shop All Novus Biologicals Products

Description
The Human Collagen X ELISA Kit (Chemiluminescence) measures human Collagen X in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 12.50-800 pg/mL |
| Assay Sensitivity | 7.50 pg/mL |
| Cross Reactivity | This kit recognizes Human COL10 in samples. No significant cross-reactivity or interference between Human COL10 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Collagen X |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?