Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Collagen XXV alpha 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7066038
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4317SP 25 μg
MAB4317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4317SP Supplier R&D Systems Supplier No. MAB4317SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Collagen XXV alpha 1 Monoclonal specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Collagen XXV alpha 1
Applications Immunohistochemistry
Classification Monoclonal
Clone 540802
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_942014
Gene Alias Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, COL25A1, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component
Gene Symbols COL25A1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Collagen XXV alpha 1 Gly189-Lys654 Accession # NP_942014
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84570
Test Specificity Detects human Collagen XXV alpha 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) COL1A1, rhCOL3A1, rhCOL4A1, rhCOL13A1, rhCOL23A1 and recombinant mouse COL13A1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.