Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Collectin-12/CL-P1 ELISA Kit

Catalog No. EH121RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH121RB 96 Tests
EH121RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH121RB Supplier Invitrogen™ Supplier No. EH121RB
Add to Cart
Add to Cart

ELISA

Human Collectin-12/CL-P1 quantitates human Collectin-12/CL-P1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Collectin-12/CL-P1.

This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Clp1; CL-P1; Colec12; collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; SCARA4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; SRCL
Assay Range 0.22 to 50 ng/mL
Assay Sensitivity 0.22 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 81035
Gene Symbol Colec12
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target COLEC12
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.