Learn More
R&D Systems™ Human Complement Factor D Magnetic Luminex™ Performance Assay
Shop All Novus Biologicals Products

Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Gene ID (Entrez) | 1675 |
| Product Line | R&D Systems |
| Synonym | Adipsin, ADN, ADNcomplement factor D, AMBP-1, C3 convertase activator, CFD, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.