Learn More
Creative Biomart Mirococept, Human Complement Receptor 1 (CR1)
Supplier:  Creative Biomart THP0193
Encompass_Preferred
The product is a truncated form of the human Complement Receptor 1 (CR1) linked to a unique Prodaptin™ construct that regulates the over-production of complement at the cell surface. It consists of the first three short consensus domains of human complement receptor 1 (CR1), manufactured in recombinant bacteria and modified with a membrane-targeting amphiphilic peptide based on the naturally occurring membrane-bound myristoyl-electrostatic switch peptide.
Catalog No. 50-208-4244
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.