Learn More
STEMCELL Technologies Human Cord Blood Plasma, Frozen, Size: 50 mL
SDP
Supplier:  STEMCELL Technologies 70020
Encompass_Preferred
Human plasma has been isolated from umbilical cord blood (CB) using centrifugation. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant.Plasma was collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. 50-197-5789
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.