Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Corin Quantikine ELISA Kit
SDP

Catalog No. DCRN00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DCRN00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DCRN00 Supplier R&D Systems™ Supplier No. DCRN00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Corin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Corin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Corin in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).

Specifications

Assay Range 75.00 to 4800 pg/mL
Assay Sensitivity 23.7 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Human Corin
For Use With (Application) ELISA
Gene ID (Entrez) 10699
Gene Symbol CORIN
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ATC2, atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Target Corin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.