Learn More
Enzo Life Sciences Human Cot DNA (500µg)
SDP
Supplier:  Enzo Life Sciences ENZGEN1160500
Encompass_Preferred
Human Cot DNA blocks repetitive sequences of DNA to suppress the level of cross-hybridization as well as non-specific binding to array surfaces. Utilizing Human Cot DNA in hybridizations increases overall sensitivity and improves experimental results. Alternative name: C0t DNA. Concentration: 1 μg/μL. Quantity: 500 μL. Formulation: Liquid. In 10 mM TrisHCl, pH 7.5, containing 1 mM EDTA. Long Term Storage: -20°C.
Catalog No. 50-201-3749
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.