Learn More
Sino Biological Human COX4I1 PAb 100UL 106894-T40-100
Supplier:  Sino Biological 106894T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human COX IV/COX4I1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1192
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.