Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human COX4I2 Full-length ORF (NP_115998.2, 1 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89940280 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-280 25 μg
89-940-279 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-940-280 Supplier Abnova™ Supplier No. H00084701P02L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes isoform 2 of subunit IV. Isoform 1 of subunit IV is encoded by a different gene, however, the two genes show a similar structural organization. Subunit IV is the largest nuclear encoded subunit which plays a pivotal role in COX regulation. [provided by RefSeq]

Sequence: MLPRAAWSLVLRKGGGGRRGMHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKTVMGCVFFFIGFAALVIWWQRVYVFPPKPITLTDERKAQQLQRMLDMKVNPVQGLASRWDYEKKQWKK

Specifications

Accession Number NP_115998.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84701
Molecular Weight (g/mol) 46.4kDa
Name COX4I2 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLPRAAWSLVLRKGGGGRRGMHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKTVMGCVFFFIGFAALVIWWQRVYVFPPKPITLTDERKAQQLQRMLDMKVNPVQGLASRWDYEKKQWKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias COX4/COX4-2/COX4B/COX4L2/COXIV-2/dJ857M17.2
Common Name COX4I2
Gene Symbol COX4I2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.