Learn More
ATCC Human Coxsackievirus A 10 (VR-168DQ) - Frozen research viral stock propagated in RD cells
Supplier: ATCC VR168DQ
Human Coxsackievirus A 10 (strain M.K., Kowalik) is supplied as a frozen research stock intended for laboratory use. The isolate was obtained from the stool of a 7-year-old child in New York in 1950 and is propagated in RD cells for virology research.
- Strain: Human Coxsackievirus A 10 (M.K., Kowalik).
- Host / propagation cell line: RD (CCL-136).
- Observed effects: Cytopathic effect, cell rounding, and cell sloughing.
- Incubation conditions: 37°C for 2-5 days.
- Format: Frozen research stock.
- Storage: -70°C or colder.
- Genome reference: GenBank AJ295191 (partial polyprotein, isolate VR-168).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.