Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CPM Full-length ORF (NP_001005502.1, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89940349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-349 10 ug
89-940-350 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-940-349 Supplier Abnova™ Supplier No. H00001368P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a membrane-bound arginine/lysine carboxypeptidase. Its expression is associated with monocyte to macrophage differentiation. This encoded protein contains hydrophobic regions at the amino and carboxy termini and has 6 potential asparagine-linked glycosylation sites. The active site residues of carboxypeptidases A and B are conserved in this protein. Three alternatively spliced transcript variants encoding the same protein have been described for this gene. [provided by RefSeq]

Sequence: MDFPCLWLGLLLPLVAALDFNYHRQEGMEAFLKTVAQNYSSVTHLHSIGKSVKGRNLWVLVVGRFPKEHRIGIPEFKYVANMHGDETVGRELLLHLIDYLVTSDGKDPEITNLINSTRIHIMPSMNPDGFEAVKKPDCYYSIGRENYNQYDLNRNFPDAFEYNNVSRQPETVAVMKWLKTETFVLSANLHGGALVASYPFDNGVQATGALYSRSLTPDDDVFQYLAHTYASRNPNMKKGDECKNKMNFPNGVTNGYSWYPLQGGMQDYNYIWAQCFEITLELSCCKYPREEKLPSFWNNNKASLIEYIKQVHLGVKGQVFDQNGNPLPNVIVEVQDRKHICPYRTNKYGEYYLLLLPGSYIINVTVPGHDPHITKVIIPEKSQNFSALKKDILLPFQGQLDSIPVSNPSCPMIPLYRNLPDHSAATKPSLFLFLVSLLHIFFK

Specifications

Accession Number NP_001005502.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1368
Molecular Weight (g/mol) 76.9kDa
Name CPM (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MDFPCLWLGLLLPLVAALDFNYHRQEGMEAFLKTVAQNYSSVTHLHSIGKSVKGRNLWVLVVGRFPKEHRIGIPEFKYVANMHGDETVGRELLLHLIDYLVTSDGKDPEITNLINSTRIHIMPSMNPDGFEAVKKPDCYYSIGRENYNQYDLNRNFPDAFEYNNVSRQPETVAVMKWLKTETFVLSANLHGGALVASYPFDNGVQATGALYSRSLTPDDDVFQYLAHTYASRNPNMKKGDECKNKMNFPNGVTNGYSWYPLQGGMQDYNYIWAQCFEITLELSCCKYPREEKLPSFWNNNKASLIEYIKQVHLGVKGQVFDQNGNPLPNVIVEVQDRKHICPYRTNKYGEYYLLLLPGSYIINVTVPGHDPHITKVIIPEKSQNFSALKKDILLPFQGQLDSIPVSNPSCPMIPLYRNLPDHSAATKPSLFLFLVSLLHIFFK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name CPM
Gene Symbol CPM
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.