Learn More
Invitrogen™ Human CRMP1 Synthetic Peptide
Description
CRMP1 Synthetic Peptide, PEP-0292, from Invitrogen.
Specifications
Specifications
| Gene ID (Entrez) | 1400 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q14194 |
| Common Name | Human DRP1 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.