Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Crossveinless-2/CV-2/BMPER ELISA Kit

Catalog No. EH132RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH132RBX5 5 x 96 Tests
EH132RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH132RBX5 Supplier Invitrogen™ Supplier No. EH132RBX5
Add to Cart
Add to Cart

ELISA

Human Crossveinless-2/CV-2/BMPER quantitates human Crossveinless-2/CV-2/BMPER in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Crossveinless-2/CV-2/BMPER.

This gene encodes a secreted protein that interacts with, and inhibits bone morphogenetic protein (BMP) function. It has been shown to inhibit BMP2- and BMP4-dependent osteoblast differentiation and BMP-dependent differentiation of the chondrogenic cells. Mutations in this gene are associated with a lethal skeletal disorder, diaphanospondylodysostosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 3110056H04Rik; BMP binding endothelial regulator; BMP-binding endothelial regulator; BMP-binding endothelial regulator precursor protein; BMP-binding endothelial regulator protein; BMPER; bone morphogenetic protein-binding endothelial cell precursor-derived regulator; Crim3; crossveinless 2; crossveinless-2; Cv2; CV-2; hCV2; KIAA1965; mCV2; Protein crossveinless-2; RGD1563373
Assay Range 0.410 to 100 ng/mL
Assay Sensitivity 0.45 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 168667
Gene Symbol Bmper
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target BMPER
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.