Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CRYAA Full-length ORF (NP_000385.1, 1 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89940614 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-614 10 ug
89-940-615 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-940-614 Supplier Abnova™ Supplier No. H00001409P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Crystallins are separated into 2 classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes major pr. of vertebrate eye lens and maintains transparency and refractive index of lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into a, b, and g families; b and g crystallins are considered as a superfamily. a and b families are further divided into acidic and basic groups. 7 protein regions exist in crystallins: 4 homologous motifs, a connecting peptide, and N- and C-terminal extensions. a crystallins are composed of 2 gene products: a-A and a-B, for acidic and basic, respectively. a crystallins can be induced by heat shock and are members of the sHSP (HSP20) family. They act as molecular chaperones although they do not renature proteins and release them in fashion of a true chaperone instead they hold them in large soluble aggregates. Post-translational modifications decrease ability to chaperone. These heterogeneous aggregates consist of 30-40 subunits; the a-A and a-B subunits have a 3:1 ratio, resp. Two additional functions of a crystallins are an autokinase activity and participation in intracellular architecture. a-A and a-B gene products are differentially expressed a-A is preferentially restricted to lens and a-B is expressed widely in many tissues and organs. Defects in this gene cause ADCC.

Sequence: MDVTIQHPWFKRTLGPFYPSRLFDQFFGEGLFEYDLLPFLSSTISPYYRQSLFRTVLDSGISEVRSDRDKFVIFLDVKHFSPEDLTVKVQDDFVEIHGKHNERQDDHGYISREFHRRYRLPSNVDQSALSCSLSADGMLTFCGPKIQTGLDATHAERAIPVSREEKPTSAPSS

Specifications

Accession Number NP_000385.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1409
Molecular Weight (g/mol) 46.3kDa
Name CRYAA (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MDVTIQHPWFKRTLGPFYPSRLFDQFFGEGLFEYDLLPFLSSTISPYYRQSLFRTVLDSGISEVRSDRDKFVIFLDVKHFSPEDLTVKVQDDFVEIHGKHNERQDDHGYISREFHRRYRLPSNVDQSALSCSLSADGMLTFCGPKIQTGLDATHAERAIPVSREEKPTSAPSS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CRYA1/HSPB4
Common Name CRYAA
Gene Symbol CRYAA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.