Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CSF1 (159 a.a.) Full-length Recombinant Protein with His tag expressed in Barley grain

Catalog No. 89940687 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-687 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-940-687 Supplier Abnova™ Supplier No. P5818
Add to Cart
Add to Cart

Used for Func, SDS-PAGE, WB-Re

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of macrophages. The active form of the protein is found extracellularly as a disulfide-linked homodimer, and is thought to be produced by proteolytic cleavage of membrane-bound precursors. The encoded protein may be involved in development of the placenta. Four transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Accession Number BAD92189
Concentration 100μg/mL
For Use With (Application) Functional Assay, SDS-PAGE, Western Blot
Formulation Lyophilized from 5mM TrisHCl, 80mM Arginine, pH 8.0
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 21kDa
Name Human CSF1 (159 a.a.) Full-length Recombinant Protein with His tag expressed in Barley grain
Purification Method Chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 1 mg
Storage Requirements Store at −20°C on dry atmosphere.After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at −20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA).Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.005ng/μg protein (0.05EU/μg) as measured by turbidimetric kinetic assay
Gene Alias MCSF, MGC31930
Common Name CSF1
Gene Symbol CSF1
Biological Activity Bioactivity is assayed by measuring the dose dependent effect of human MCSF on proliferation of M-NFS-60 cells. The ED50 lies typically in the range of 0.51-0.76ng/mL.
Species Plants
Recombinant Recombinant
Protein Tag His Tag
Expression System Barley grain (Hordeum vulgare) expression system
Form Lyophilized
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.