Learn More
ACROBiosystems Human CSPG4, His Tag (CS4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Ser 2224 (Accession # Q6UVK1-1).
Supplier:  ACROBiosystems CS4H52H3100UG
Encompass_Preferred
Chondroitin sulfate proteoglycan 4,also as knowed melanoma-associated chondroitin sulfate proteoglycan (MCSP) is a chondroitin sulfate proteoglycan that in humans is encoded by the CSPG4 gene. CSPG4 plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. It represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.
Catalog No. 50-225-3858
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.