Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CTACK (CCL27) Recombinant Protein, PeproTech®

Catalog No. 30054250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30054250UG 250 μg
30054100UG 100 μg
300-54-1MG 1 mg
300-54-20UG 20 μg
30054500UG 500 μg
300-54-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30054250UG Supplier Gibco™ Supplier No. 30054250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human CTACK is a 10.2 kDa protein containing 88 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL27 gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The protein encoded by this gene is chemotactic for skin-associated memory T lymphocytes. This cytokine may also play a role in mediating homing of lymphocytes to cutaneous sites. It specifically binds to chemokine receptor 10 (CCR10). Studies of a similar murine protein indicate that these protein-receptor interactions have a pivotal role in T cell-mediated skin inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y4X3
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 10850
Molecular Weight (g/mol) 10.2 kDa
Name Human CTACK (CCL27)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias ALP; AW558992; CC chemokine ILC; C-C motif chemokine 27; C-C motif chemokine ligand 27; Ccl27; Ccl27a; chemokine (C-C motif) ligand 27; chemokine (C-C motif) ligand 27 A; CTACK; CTAK; cutaneous T-cell attracting chemokine; cutaneous T-cell-attracting chemokine; ESkine; IL-11 R-alpha-locus chemokine; IL-11 Ralpha-locus chemokine; ILC; PESKY; SCYA27; Scya27a; Skinkine; skinskine; small inducible cytokine A27; small inducible cytokine A27a; small inducible cytokine subfamily A (Cys-Cys), member 27; small-inducible cytokine A27
Common Name CCL27
Gene Symbol CCL27
Biological Activity Determined by its ability to chemoattract CXCR3 transfected cells using a concentration of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence FLLPPSTACC TQLYRKPLSD KLLRKVIQVE LQEADGDCHL QAFVLHLAQR SICIHPQNPS LSQWFEHQER KLHGTLPKLN FGMLRKMG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.