Learn More
ACROBiosystems ACROBiosystems Human CTGF / CCN2 Protein, His Tag
Supplier:  ACROBiosystems CTFH52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CTGF, His Tag (CTF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # Q5M8T4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.4 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CTGF, His Tag (Cat. No. CTF-H52H5) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CTGF Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-210-6617