Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CTGFL/WISP-2 Recombinant Protein, PeproTech®

Catalog No. 120165UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-16-5UG 5 μg
12016100UG 100 μg
120-16-1MG 1 mg
12016250UG 250 μg
12016500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 120-16-5UG Supplier Gibco™ Supplier No. 120165UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human CTGFL/WISP-2 is a 24.3 kDa protein consisting of 228 amino acid residues. Mature human CTGFL/WISP-2 is a 24.8 kDa polypeptide protein containing 227 amino acids. It is composed of 3 distinct domains; the IGF-Binding Protein domain (IGF-BP), the Thrombospondin type I repeat (TSP type I), and von Willebrand Factor C motif (VWFC). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Wisp2 a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like (CT) domain. Wisp2 lacks the CT domain which is implicated in dimerization and heparin binding. It is 72% identical to the mouse protein at the amino acid level. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. Its expression in colon tumors is reduced while the other two WISP members are overexpressed in colon tumors. It is expressed at high levels in bone tissue, and may play an important role in modulating bone turnover.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O76076
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 8839
Molecular Weight (g/mol) 24.3 kDa
Name Human CTGFL/WISP-2
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CCN family member 5; CCN family protein COP-1; Ccn5; connective tissue growth factor-like protein; Connective tissue growth factor-related protein 58; Cop1; Crgr4; CT58; CTGFL; CTGF-L; Rcop1; UNQ228/PRO261; WISP2; WISP-2; WNT1 inducible signaling pathway protein 2; WNT1-inducible-signaling pathway protein 2
Common Name WISP2
Gene Symbol CCN5
Biological Activity The ED50 was determined by its ability to inhibit IGF-II induced proliferation of MCF-7 is between 10-20 ng/ml in the presence of 15 ng/ml of human IGF-II.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.