Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CTLA-4 Fc Recombinant Protein, PeproTech®

Catalog No. 310051MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
200 μg
50 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310-05-1MG 1 mg
31005200UG 200 μg
31005500UG 500 μg
310-05-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 310-05-1MG Supplier Gibco™ Supplier No. 310051MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human CTLA-4 Fc is a glycosylated, disulfide-linked homodimer of 714 amino acid residues whose monomer consists of the 124-amino-acid length extracellular portion of CTLA-4 fused to the 231-amino-acid length Fc portion of human IgG1 by two glycines. The calculated molecular weight of Recombinant Human CTLA-4 Fc dimer is 78.7 kDa; however, due to glycosylation, the monomer and dimer migrate at apparent molecular weights of approximately 45-50 kDa and 80-90 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity than CD28. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhbits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16410
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 1493
Molecular Weight (g/mol) 78.7 kDa
Name Human CTLA-4 Fc
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form
Common Name CTLA-4 (CD152)
Gene Symbol CTLA4
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence MHVAQPAVVL ASSRGIASFV CEYASPGKAT EVRVTVLRQA DSQVTEVCAA TYMMGNELTF LDDSICTGTS SGNQVNLTIQ GLRAMDTGLY ICKVELMYPP PYYLGIGNGT QIYVIDPEPC PDSDGGPKSC DKTHTCPPCP APELLGGPSV FLFPPKPKDT LMISRTPEVT CVVVDVSHED PEVKFNWYVD GVEVHNAKTK PREEQYNSTY RVVSVLTVLH QDWLNGKEYK CKVSNKALPA PIEKTISKAK GQPREPQVYT LPPSRDELTK NQVSLTCLVK GFYPSDIAVE WESNGQPENN YKTTPPVLDS DGSFFLYSKL TVDKSRWQQG NVFSCSVMHE ALHNHYTQKS LSLSPGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.