Learn More
Sino Biological Recombinant Human beta-Catenin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11279H07E100
Encompass_Preferred
A DNA sequence encoding the Human CTNNB1 (P35222-1) (Asn138-Leu781) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-4741
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.