Learn More
Description
Specifications
Specifications
| Assay Range | 78.10 to 5,000pg/mL |
| Conjugate | Biotin |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sufficient For | Five 96-well plates |
| For Use With (Application) | ELISA |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
