Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CTRP5/C1qTNF5 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3167
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3167 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3167 Supplier R&D Systems Supplier No. BAF3167
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

C1qTNF5/CTRP5 Polyclonal specifically detects C1qTNF5/CTRP5 in Human samples. It is validated for Western Blot.

Specifications

Antigen C1qTNF5/CTRP5
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9BXJ0
Gene Alias C1q and tumor necrosis factor related protein 5, C1qTNF5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3
Gene Symbols C1QTNF5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CTRP5/C1qTNF5 Ser16-Ala243 Accession # Q9BXJ0
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114902
Test Specificity Detects human CTRP5/C1qTNF5 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) CTRP4 is observed and less than 1% cross-reactivity with rhCTRP1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.