Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CUL4B Full-length ORF (AAH36216.1, 1 a.a. - 913 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89940995 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-995 25 μg
89-940-994 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-940-995 Supplier Abnova™ Supplier No. H00008450P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the cullin family. The encoded protein forms a complex that functions as an E3 ubiquitin ligase and catalyzes the polyubiquitination of specific protein substrates in the cell. The protein interacts with a ring finger protein, and is required for the proteolysis of several regulators of DNA replication including chromatin licensing and DNA replication factor 1 and cyclin E. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MMSQSSGSGDGNDDEATTSKDGGFSSPSPSAAAAAQEVRSATDGNTSTTPPTSAKKRKLNSSSSSSSNSSNEREDFDSTSSSSSTPPLQPRDSASPSTSSFCLGVSVAASSHVPIQKKLRFEDTLEFVGFDAKMAEESSSSSSSSSPTAATSQQQQLKNKSILISSVASVHHANGLAKSSTTVSSFANSKPGSAKKLVIKNFKDKPKLPENYTDETWQKLKEAVEAIQNSTSIKYNLEELYQAVENLCSYKISANLYKQLRQICEDHIKAQIHQFREDSLDSVLFLKKIDRCWQNHCRQMIMIRSIFLFLDRTYVLQNSMLPSIWDMGLELFRAHIISDQKVQNKTIDGILLLIERERNGEAIDRSLLRSLLSMLSDLQIYQDSFEQRFLEETNRLYAAEGQKLMQEREVPEYLHHVNKRLEEEADRLITYLDQTTQKSLIATVEKQLLGEHLTAILQKGLNNLLDENRIQDLSLLYQLFSRVRGGVQVLLQQWIEYIKAFGSTIVINPEKDKTMVQELLDFKDKVDHIIDICFLKNEKFINAMKEAFETFINKRPNKPAELIAKYVDSKLRAGNKEATDEELEKMLDKIMIIFRFIYGKDVFEAFYKKDLAKRLLVGKSASVDAEKSMLSKLKHECGAAFTSKLEGMFKDMELSKDIMIQFKQYMQNQNVPGNIELTVNILTMGYWPTYVPMEVHLPPEMVKLQEIFKTFYLGKHSGRKLQWQSTIGHCVLKAEFKEGKKELQVSLFQTLVLLMFNEGEEFSLEEIKQATGIEDGELRRTLQSLACGKARVLAKNPKGKDIEDGDKFICNDDFKHKLFRIKINQIQMKETVEEQASTTERVFQDRQYQIDAAIVRIMKMRKTLSHNLLVSEVYNQLKFPVKPADLKKRIESLIDRDYMERDKENPNQYNYIA

Specifications

Accession Number AAH36216.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8450
Molecular Weight (g/mol) 130.4kDa
Name CUL4B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MMSQSSGSGDGNDDEATTSKDGGFSSPSPSAAAAAQEVRSATDGNTSTTPPTSAKKRKLNSSSSSSSNSSNEREDFDSTSSSSSTPPLQPRDSASPSTSSFCLGVSVAASSHVPIQKKLRFEDTLEFVGFDAKMAEESSSSSSSSSPTAATSQQQQLKNKSILISSVASVHHANGLAKSSTTVSSFANSKPGSAKKLVIKNFKDKPKLPENYTDETWQKLKEAVEAIQNSTSIKYNLEELYQAVENLCSYKISANLYKQLRQICEDHIKAQIHQFREDSLDSVLFLKKIDRCWQNHCRQMIMIRSIFLFLDRTYVLQNSMLPSIWDMGLELFRAHIISDQKVQNKTIDGILLLIERERNGEAIDRSLLRSLLSMLSDLQIYQDSFEQRFLEETNRLYAAEGQKLMQEREVPEYLHHVNKRLEEEADRLITYLDQTTQKSLIATVEKQLLGEHLTAILQKGLNNLLDENRIQDLSLLYQLFSRVRGGVQVLLQQWIEYIKAFGSTIVINPEKDKTMVQELLDFKDKVDHIIDICFLKNEKFINAMKEAFETFINKRPNKPAELIAKYVDSKLRAGNKEATDEELEKMLDKIMIIFRFIYGKDVFEAFYKKDLAKRLLVGKSASVDAEKSMLSKLKHECGAAFTSKLEGMFKDMELSKDIMIQFKQYMQNQNVPGNIELTVNILTMGYWPTYVPMEVHLPPEMVKLQEIFKTFYLGKHSGRKLQWQSTIGHCVLKAEFKEGKKELQVSLFQTLVLLMFNEGEEFSLEEIKQATGIEDGELRRTLQSLACGKARVLAKNPKGKDIEDGDKFICNDDFKHKLFRIKINQIQMKETVEEQASTTERVFQDRQYQIDAAIVRIMKMRKTLSHNLLVSEVYNQLKFPVKPADLKKRIESLIDRDYMERDKENPNQYNYIA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686F1470/KIAA0695/MRXHF2/MRXSC/SFM2
Common Name CUL4B
Gene Symbol CUL4B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.