Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CXCL-17 ELISA Kit

Catalog No. EH137RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH137RBX10 10 x 96 Tests
EH137RBX5 5 x 96 Tests
EH137RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH137RBX10 Supplier Invitrogen™ Supplier No. EH137RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human CXCL-17 quantitates human CXCL-17 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CXCL-17.

CXCL17 is a chemokine that acts as chemoattractant for monocytes, macrophages and dendritic cells. In particular, CXCL17 is a mucosal chemokine that attracts dendritic cells and blood monocytes to the lungs. CXCL17 plays a role in angiogenesis and possibly in the development of tumors. Further, CXCL17 acts as an anti-inflammatory in the stomach and may play a role in the innate defense against infections. CXCL17 also activates the C-X-C chemokine receptor GPR35 to induce a rapid and transient rise in the level of intracellular calcium ions. 4-Cys CXCL17 seems to exhibit much higher chemoattractant potency on monocytes and macrophages than 6-Cys CXCL17. Diseases associated with CXCL17 dysfunction include cholera.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 13.6 kDa protein; 13.6-kDa protein; 4-Cys CXCL17; 6-Cys CXCL17; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 17; CXC; CXC motif chemokine; C-X-C motif chemokine 17; C-X-C motif chemokine ligand 17; CXCL; Cxcl17; Dcip1; Dendritic cell and monocyte chemokine-like protein; DMC; UNQ473; UNQ473/PRO842; VCC1; VCC-1; VEGF coregulated chemokine 1; VEGF co-regulated chemokine 1
Assay Range 0.6 to 150 ng/mL
Assay Sensitivity 0.6 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 284340
Gene Symbol CXCL17
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CXCL17
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.