Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CXCL1/GRO alpha Quantikine ELISA Kit
SDP

Catalog No. SGR00B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SGR00B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SGR00B Supplier R&D Systems™ Supplier No. SGR00B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human CXCL1/GRO alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CXCL1/GRO alpha Quantikine ELISA Kit quantifies human CXCL1/GRO alpha/KC/CINC-1 in cell culture supernates (200 ul), serum (200 ul), platelet-poor edta plasma (200 ul), platelet-poor heparin plasma (200 ul), platelet-poor citrate plasma (200 ul).

Specifications

Assay Range 31.30 - 1000pg/mL
Assay Sensitivity 10pg/mL
Product Type Human CXCL1/GRO alpha Quantikine ELISA Kit, 2nd Gen
Sample Type Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human GRO alpha
Gene ID (Entrez) 2919
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (200uL),Serum (200uL),Platelet-poor EDTA Plasma (200uL),Platelet-poor Heparin Plasma (200uL),Platelet-poor Citrate Plasma (200uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Target CXCL1/GRO alpha/KC/CINC-1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.